5-Fluoroindole ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
5-Fluoro-1H-indole
F0369-1G
F0369-5G
TCF0369-001GEA
35
USD
TCF0369-001G
TCF0369-005G
5-Fluoroindole ≥98.0%
5-Fluoroindole
CAS Number: 399-52-0
MDL Number: MFCD00005671
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method:
Formula:
C₈H₆FN Boiling Pt: 120 °C Melting Pt: 47 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005671 CAS Number: 399-52-0 EINECS: 206-917-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...