3-Fluoro-o-xylene ≥98.0%

Supplier: TCI America

Synonyms: 1-Fluoro-2,3-dimethylbenzene, 1,2-Dimethyl-3-fluorobenzene

F0332-5ML
TCF0332-5MLEA 43.9 USD
TCF0332-5ML
3-Fluoro-o-xylene ≥98.0%
3-Fluoro-o-xylene

CAS Number: 443-82-3 MDL Number: MFCD00000323 Molecular Formula: C8H9F Molecular Weight: 124.16 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 148 Flash Point (°C): 36 Specific Gravity (20/20): 1.01


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: 2,3-Dimethylfluorobenzene, 1-Fluoro-2,3-dimethylbenzene

Formula: C₈H₉F
MW: 124.16 g/mol
Boiling Pt: 148 °C
Density: 1.01 (20 °C)
Flash Pt: 36 °C
MDL Number: MFCD00000323
CAS Number: 443-82-3
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=235f0608374dae4bc9af0262f5c5513cb5f898c5&vid=235f0608374dae4bc9af0262f5c5513cb5f898c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR