2-Fluoro-α,α,α-trichlorotoluene ≥98.0%

Supplier: TCI America

F0290-5G
TCF0290-025GEA 128.76 USD
TCF0290-025G TCF0290-5G
2-Fluoro-α,α,α-trichlorotoluene ≥98.0%
2-Fluoro-α,α,α-trichlorotoluene

CAS Number: 488-98-2 MDL Number: MFCD00000787 Molecular Formula: C7H4Cl3F Molecular Weight: 213.46 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 75 Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: 2-Fluoro-alpha,alpha,alpha-trichlorotoluene

Formula: C₇H₄Cl₃F
Boiling Pt: 75 °C
Melting Pt: –20 °C
Density: 1.4523 g/cm³ (18.6 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000787
CAS Number: 488-98-2
EINECS: 207-692-5
UN: 2810
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72c2b49596cdc8ae12440b5078e9208bd8a97f1f&vid=72c2b49596cdc8ae12440b5078e9208bd8a97f1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR