3'-Fluoroacetophenone ≥97.0% (by GC)

Supplier: TCI America

F0262-25G F0262-5G
TCF0262-25GEA 169 USD
TCF0262-25G TCF0262-5G
3'-Fluoroacetophenone ≥97.0% (by GC)
3'-Fluoroacetophenone

CAS Number: 455-36-7 MDL Number: MFCD00000338 Molecular Formula: C8H7FO Molecular Weight: 138.14 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 81 Melting point (°C): -3 Flash Point (°C): 80 Specific Gravity (20/20): 1.14

Formula: C₈H₇FO
Boiling Pt: 81 °C
Melting Pt: –3 °C
Density: 1.14 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00000338
CAS Number: 455-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0dda57f76fc4a9b2a926fc15f035fb70547850&vid=ea0dda57f76fc4a9b2a926fc15f035fb70547850&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR