3'-Fluoroacetophenone ≥97.0% (by GC)
Supplier: TCI America
CAS Number: 455-36-7
MDL Number: MFCD00000338
Molecular Formula: C8H7FO
Molecular Weight: 138.14
Purity/Analysis Method:
Formula:
C₈H₇FO Boiling Pt: 81 °C Melting Pt: –3 °C Density: 1.14 (20 °C) Flash Pt: 80 °C |
MDL Number:
MFCD00000338 CAS Number: 455-36-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...