9-Fluorenylmethanol ≥98.0%
Supplier: TCI America
Synonyms:
(Fluoren-9-yl)methanol, 9-Fluorenmethanol, 9-Fluorenemethanol
F0200-25G
F0200-5G
TCF0200-25GEA
53
USD
TCF0200-25G
TCF0200-005G
9-Fluorenylmethanol ≥98.0%
9-Fluorenylmethanol
CAS Number: 24324-17-2
MDL Number: MFCD00001139
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method:
Other Product Names / Synonyms: 9-Hydroxymethylfluorene
Formula:
C₁₄H₁₂O Boiling Pt: 337 °C (1013 hPa) Melting Pt: 105 °C |
MDL Number:
MFCD00001139 CAS Number: 24324-17-2 EINECS: 246-167-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...