2-Fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

F0195-500G F0195-25G F0195-100G
TCF0195-500GEA 153 USD
TCF0195-500G TCF0195-25G TCF0195-100G
2-Fluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
2-Fluorobenzoyl chloride

CAS Number: 393-52-2 MDL Number: MFCD00000656 Molecular Formula: C7H4ClFO Molecular Weight: 158.56 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 189 Flash Point (°C): 82 Specific Gravity (20/20): 1.34

Formula: C₇H₄ClFO
Boiling Pt: 189 °C
Density: 1.34 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e31175a4a574e00c34f80bcc90aeebadcc7cd544&vid=e31175a4a574e00c34f80bcc90aeebadcc7cd544&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR