4-Fluoro-3-nitroaniline ≥98.0%

Supplier: TCI America
Warning

F0168-25G
TCF0168-010GEA 25.53 USD
TCF0168-010G TCF0168-025G
4-Fluoro-3-nitroaniline ≥98.0%
4-Fluoro-3-nitroaniline

CAS Number: 364-76-1 MDL Number: MFCD00007833 Molecular Formula: C6H5FN2O2 Molecular Weight: 156.12 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 97 Flash Point (°C): 91

Formula: C₆H₅FN₂O₂
Melting Pt: 97 °C
Density: 0.43 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007833
CAS Number: 364-76-1
EINECS: 206-665-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bec1d4aadd2f52a5caa7f99c658f930ab2aaa59&vid=e692d350300f0ad292d6fab1c9cc2563b47a41bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR