2-Fluorophenol ≥98.0%

Supplier: TCI America
Danger

Synonyms: o-Fluorophenol

F0150-250G F0150-25G
TCF0150-250GEA 102.74 USD
TCF0150-250G TCF0150-25G
2-Fluorophenol ≥98.0%
2-Fluorophenol

CAS Number: 367-12-4 MDL Number: MFCD00002155 Molecular Formula: C6H5FO Molecular Weight: 112.10 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 152 Flash Point (°C): 47 Freezing point (°C): 12 Specific Gravity (20/20): 1.22

Formula: C₆H₅FO
Boiling Pt: 152 °C
Melting Pt: 16 °C
Density: 1.22 (20 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00002155
CAS Number: 367-12-4
EINECS: 206-681-2
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3376f687c7d90f61161fddb4c9c33041b590ee46&vid=874acab580ab0140ff2a1f447c0937af3936a7a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR