Fast green FCF ≥85.0% (by HPLC and UV method)

Supplier: TCI America
Warning

F0099-1G F0099-10G
TCF0099-001GEA 26.87 USD
TCF0099-001G TCF0099-010G
Fast green FCF ≥85.0% (by HPLC and UV method)
Fast Green FCF

CAS Number: 2353-45-9 MDL Number: MFCD00013053 Molecular Formula: C37H36N2O10S3 Molecular Weight: 808.84 Purity/Analysis Method: 85.0% (HPLC,E) Form: Crystal Lambda max.: 624 nm (0.02mol/L AcONH4 sol.)

Formula: C₃₇H₃₄N₂Na₂O₁₀S₃
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00013053
CAS Number: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff2ef5c8bd90972315d424c05b8aab707fa7e609&vid=ff2ef5c8bd90972315d424c05b8aab707fa7e609&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (HPLC,E)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR