Fluorescein disodium salt
Supplier: TCI America
F0096-25G
F0096-500G
F0096-100G
TCF0096-025GEA
27.97
USD
TCF0096-025G
TCF0096-500G
TCF0096-100G
Fluorescein disodium salt
Fluorescein disodium salt
CAS Number: 518-47-8
MDL Number: MFCD00167039
Molecular Formula: C20H12O5
Molecular Weight: 376.27
Form: Crystal
Color: Deep Yellow
Other Product Names / Synonyms: Fluorescein Sodium Salt
Formula:
C₂₀H₁₀Na₂O₅ Melting Pt: > 300 °C Density: 1.601 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167039 CAS Number: 518-47-8 EINECS: 208-253-0 Merck Index: 13,04185 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...