Fluorescein disodium salt

Supplier: TCI America

F0096-25G F0096-500G F0096-100G
TCF0096-025GEA 27.97 USD
TCF0096-025G TCF0096-500G TCF0096-100G
Fluorescein disodium salt
Fluorescein disodium salt

CAS Number: 518-47-8
MDL Number: MFCD00167039
Molecular Formula: C20H12O5
Molecular Weight: 376.27
Form: Crystal
Color: Deep Yellow


Other Product Names / Synonyms: Fluorescein Sodium Salt

Formula: C₂₀H₁₀Na₂O₅
Melting Pt: > 300 °C
Density: 1.601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e0f87df032e8949e48b76f7e79b205d17cc9a93&vid=dd2dc9e704eeef0d55f25d2692d323af2c03acba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR