Ethyl formate ≥98.0%

Supplier: TCI America
Danger

Synonyms: Formic acid ethyl ester

F0085-100ML
TCF0085-100MLEA 100 USD
TCF0085-100ML
Ethyl formate ≥98.0%
Ethyl formate

[for Spectrophotometry] CAS Number: 109-94-4 MDL Number: MFCD00003294 Molecular Formula: C3H6O2 Molecular Weight: 74.08 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 54 Melting point (°C): -80 Flash Point (°C): -20 Specific Gravity (20/20): 0.92


Product Categories: Analytical Science, Solvents for Spectrophotometry Other Product Names / Synonyms: Formic Acid Ethyl Ester

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 54 °C
Melting Pt: –80 °C
Density: 0.92 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: 3,II
Merck Index: 13,03841
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdd26bc3cf055e6116f68db38d215e830edeb04a&vid=6805f2b7c1009a657b3f5c3c56a314f6b0629e68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR