Fumaric acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Warning

F0067-25G F0067-500G
TCF0067-025GEA 26.9 USD
TCF0067-025G TCF0067-500G
Fumaric acid ≥99.0% (by GC, titration analysis)
Fumaric acid

CAS Number: 110-17-8 MDL Number: MFCD00002700 Molecular Formula: C4H4O4 Molecular Weight: 116.07 Purity/Analysis Method: 99.0% (GC,T) Form: Crystal Melting point (°C): 287


Other Product Names / Synonyms: trans-2-Butenedioic Acid

Formula: C₄H₄O₄
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 287 °C
Density: 1.625 g/cm³ (20 °C)
Flash Pt: 273 °C
MDL Number: MFCD00002700
CAS Number: 110-17-8
EINECS: 203-743-0
Merck Index: 13,04308
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e21f7e9afb6b200e1efd8a975187627f1ea2bf00&vid=e21f7e9afb6b200e1efd8a975187627f1ea2bf00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR