Folic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
F0043-25G
TCF0043-025GEA
93.07
USD
TCF0043-025G
Folic acid ≥98.0% (by HPLC, titration analysis)
Folic acid
CAS Number: 59-30-3
MDL Number: MFCD00079305
Molecular Formula: C19H19N7O6
Molecular Weight: 441.40
Purity/Analysis Method:
Product Categories: Bioscience, Vitamins
Other Product Names / Synonyms: Vitamin B9, Vitamin M, Pteroyl-L-glutamic Acid, PteGlu
Formula:
C₁₉H₁₉N₇O₆·H₂O MW: 441.41 g/mol |
MDL Number:
MFCD00079305 CAS Number: 59-30-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...