Rose Bengal

Supplier: TCI America

F0001-25G
TCF0001-025GEA 136 USD
TCF0001-025G
Rose Bengal
Rose Bengal

CAS Number: 632-69-9
MDL Number: MFCD00151169
Molecular Weight: 1017.63
Form: Powder
Color: Red

Formula: C₂₀H₄Cl₄I₄O₅
MDL Number: MFCD00151169
CAS Number: 632-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4ea89a7d419a4b287e0d53e74559e4c2ac26f0d&vid=7130a33d35ade05667569738cb83e607da01fba6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR