Ethidium bromide 0.5mg/mL in water for electrophoresis

Supplier: TCI America


Packaged in dropper bottle.

E1363-10ML
TCE1363-10MLEA 52 USD
TCE1363-10ML
Ethidium bromide 0.5mg/mL in water for electrophoresis
Ethidium bromide
Formula: C₂₁H₂₀BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6
Merck Index: 14,04731

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
DNA staining test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR