Ethyl-4-chloroquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate

E1201-200MG E1201-1G
TCE1201-200MGEA 66 USD
TCE1201-200MG TCE1201-1G
Ethyl-4-chloroquinoline-3-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl 4-chloroquinoline-3-carboxylate
Formula: C₁₂H₁₀ClNO₂
Boiling Pt: 129 °C
MDL Number: MFCD00173407
CAS Number: 13720-94-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR