Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 641569-96-2
MDL Number: MFCD11100711
Molecular Formula: C11H15N3O2
Molecular Weight: 284.27
Purity/Analysis Method:
Other Product Names / Synonyms: 3-Carbamimidoylamino-4-methylbenzoic Acid Ethyl Ester Nitrate, Ethyl 3-Carbamimidoylamino-p-toluate Nitrate, 3-Carbamimidoylamino-p-toluic Acid Ethyl Ester Nitrate
Formula:
C₁₁H₁₅N₃O₂ Melting Pt: 199 °C |
MDL Number:
MFCD11100711 CAS Number: 641569-96-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...