Ethyl-4-phenylbutyrate ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 10031-93-3
MDL Number: MFCD00026923
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method:
Other Product Names / Synonyms: 4-Phenylbutyric Acid Ethyl Ester
Formula:
C₁₂H₁₆O₂ Boiling Pt: 100 °C Density: 1.00 (20 °C) |
MDL Number:
MFCD00026923 CAS Number: 10031-93-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...