Eplerenone ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester
CAS Number: 107724-20-9
MDL Number: MFCD07783647
Molecular Formula: C24H30O6
Molecular Weight: 414.50
Purity/Analysis Method:
Product Categories: Bioscience, Antihypertensive Ingredients, Other Steroids
Other Product Names / Synonyms: 9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester
Formula:
C₂₄H₃₀O₆ MW: 414.50 g/mol Melting Pt: 242 °C |
MDL Number:
MFCD07783647 CAS Number: 107724-20-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...