Ethyl-trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)

Supplier: TCI America

E0838-5G E0838-25G
TCE0838-5GEA 233 USD
TCE0838-5G TCE0838-25G
Ethyl-trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)
Ethyl-trans-4-hydroxycyclohexanecarboxylate

CAS Number: 3618-04-0 Molecular Formula: C9H16O3 Molecular Weight: 172.22 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 144 Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: trans-4-Hydroxycyclohexanecarboxylic Acid Ethyl Ester

Formula: C₉H₁₆O₃
Boiling Pt: 144 °C
Density: 1.06 (20 °C)
MDL Number: MFCD16295187
CAS Number: 3618-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1efef3b85e38963af450c29d7d6b6db518d1b15&vid=b1efef3b85e38963af450c29d7d6b6db518d1b15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR