1-Propyl-4-ethynylbenzene ≥96.0%
Supplier: TCI America
Synonyms:
1-Ethynyl-4-propylbenzene, (4-Propylphenyl)acetylene, 4-n-Propylphenylacetylene, (4-Propylphenyl)ethyne
E0750-5G
E0750-25G
TCE0750-5GEA
50
USD
TCE0750-5G
TCE0750-25G
1-Propyl-4-ethynylbenzene ≥96.0%
1-Propyl-4-ethynylbenzene
CAS Number: 62452-73-7
MDL Number: MFCD00173886
Molecular Formula: C11H12
Molecular Weight: 144.22
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Propylphenylacetylene
Formula:
C₁₁H₁₂ Density: 0.91 (20 °C) |
MDL Number:
MFCD00173886 CAS Number: 62452-73-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...