Etoposide ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(5R,5aR,8aR,9S)-9-(((4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one, 4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)
CAS Number: 33419-42-0
Molecular Formula: C29H32O13
Molecular Weight: 588.56
Purity/Analysis Method:
Product Categories: Bioscience, Glycosides, Glucose, Inhibitors (for Pharmacological Research), Antitumor Ingredients, Cancer Research Reagents
Other Product Names / Synonyms: 4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)
Formula:
C₂₉H₃₂O₁₃ MW: 588.56 g/mol Melting Pt: 251 °C |
MDL Number:
MFCD00869325 CAS Number: 33419-42-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...