N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine sulfate ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 25646-77-9
Molecular Formula: C11H18N2O
Molecular Weight: 292.35
Purity/Analysis Method:
Other Product Names / Synonyms: 2-[(4-Amino-3-methylphenyl)ethylamino]ethanol Sulfate
Formula:
C₁₁H₁₈N₂O Melting Pt: 155 °C |
CAS Number:
25646-77-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...