1-Fluoro-2-ethynylbenzene ≥97.0%

Supplier: TCI America

Synonyms: (2-Fluorophenyl)ethyne, 1-Ethynyl-2-fluorobenzene, (2-Fluorophenyl)acetylene

E0654-5G
TCE0654-5GEA 100 USD
TCE0654-5G
1-Fluoro-2-ethynylbenzene ≥97.0%
1-Fluoro-2-ethynylbenzene

CAS Number: 766-49-4 Molecular Formula: C8H5F Molecular Weight: 120.13 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 40 Flash Point (°C): 37 Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Terminal Alkynes (Click Chemistry) Other Product Names / Synonyms: 2-Fluorophenylacetylene

Formula: C₈H₅F
MW: 120.13
Boiling Pt: 40 °C
Density: 1.06 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00799540
CAS Number: 766-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbbc2add0ff3f3836ca0d229875d8f672aac97cc&vid=fbbc2add0ff3f3836ca0d229875d8f672aac97cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR