9-Ethynyl-9-fluorenol ≥98.0%
Supplier: TCI America
CAS Number: 13461-74-0
MDL Number: MFCD00041555
Molecular Formula: C15H10O
Molecular Weight: 206.24
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 9-Ethynyl-9-hydroxyfluorene, (9-Hydroxy-9-fluorenyl)acetylene
Formula:
C₁₅H₁₀O |
MDL Number:
MFCD00041555 CAS Number: 13461-74-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...