N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 82717-96-2
MDL Number: MFCD00209976
Molecular Formula: C15H21NO4
Molecular Weight: 279.34
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-[(S)-1-Carboxyethyl]-L-homophenylalanine Ethyl Ester
Formula:
C₁₅H₂₁NO₄ Melting Pt: 150 °C |
MDL Number:
MFCD00209976 CAS Number: 82717-96-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...