1,2-Epoxycyclopentane ≥97.0%

Supplier: TCI America

E0513-5G E0513-25G
TCE0513-005GEA 36 USD
TCE0513-005G TCE0513-25G
1,2-Epoxycyclopentane ≥97.0%
1,2-Epoxycyclopentane

CAS Number: 285-67-6 MDL Number: MFCD00005161 Molecular Formula: C5H8O Molecular Weight: 84.12 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 102 Flash Point (°C): 10 Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Cyclopentene Oxide, 6-Oxabicyclo[3.1.0]hexane

Formula: C₅H₈O
Boiling Pt: 102 °C
Density: 0.97 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00005161
CAS Number: 285-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eccad44b75bfa358ee45f1d1512c74b59aaf910d&vid=eccad44b75bfa358ee45f1d1512c74b59aaf910d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR