Eriochrome Black T

Supplier: TCI America
Warning

Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

E0214-25G
TCE0214-025GEA 81.52 USD
TCE0214-025G
Eriochrome Black T
Eriochrome Black T

CAS Number: 1787-61-7 MDL Number: MFCD00003935 Molecular Formula: C20H13N3O7S Molecular Weight: 461.38 Form: Crystal Color: Black


Other Product Names / Synonyms: 1-(1-Hydroxy-2-naphthylazo)-6-nitro-2-naphthol-4-sulfonic Acid Sodium Salt, Mordant Black 11

Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III
Merck Index: 13,03702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6f629fc77a425ef4f6dab6e125463f3dfd0c483&vid=c6f629fc77a425ef4f6dab6e125463f3dfd0c483&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR