Eugenol acetate ≥98.0%

Supplier: TCI America

Synonyms: 4-Allyl-2-methoxyphenyl acetate, Acetyleugenol

E0210-25G
TCE0210-025GEA 68.36 USD
TCE0210-025G
Eugenol acetate ≥98.0%
Eugenol acetate

CAS Number: 93-28-7 MDL Number: MFCD00026191 Molecular Formula: C12H14O3 Molecular Weight: 206.24 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 164 Flash Point (°C): 140 Freezing point (°C): 26


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals) Other Product Names / Synonyms: Acetyleugenol

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Boiling Pt: 164 °C
Flash Pt: 140 °C
MDL Number: MFCD00026191
CAS Number: 93-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=743dda9704d0a3fd948b1638674ee8198c60f88a&vid=743dda9704d0a3fd948b1638674ee8198c60f88a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR