EDTA (ethylenediamine tetraacetic acid) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

E0084-25G E0084-500G
TCE0084-025GEA 31.21 USD
TCE0084-025G TCE0084-500G
EDTA (ethylenediamine tetraacetic acid) ≥98.0% (by titrimetric analysis)
EDTA (Ethylenediamine tetraacetic acid)

CAS Number: 60-00-4 MDL Number: MFCD00003541 Molecular Formula: C10H16N2O8 Molecular Weight: 292.24 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 240


Product Categories: Analytical Science, Chelating Agents for Radiopharmaceutical Research, Complexones (Chelating Agents) Other Product Names / Synonyms: (Ethylenedinitrilo)tetraacetic Acid, EDTA

Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 240 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38d4d94247c54758ab8dcbfcc02068d4d3fc21c3&vid=38d4d94247c54758ab8dcbfcc02068d4d3fc21c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR