4-Ethylbenzenesulfonyl chloride ≥95.0%

Supplier: TCI America

Synonyms: 4-Ethylbenzene-1-sulfonyl chloride, 4-Ethylbenzene-1-sulphonyl chloride

E0065-25G
TCE0065-25GEA 99.4 USD
TCE0065-25G
4-Ethylbenzenesulfonyl chloride ≥95.0%
4-Ethylbenzenesulphonyl chloride

CAS Number: 16712-69-9 MDL Number: MFCD00041480 Molecular Formula: C8H9ClO2S Molecular Weight: 204.67 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Specific Gravity (20/20): 1.28

Formula: C₈H₉ClO₂S
Density: 1.28 (20 °C)
MDL Number: MFCD00041480
CAS Number: 16712-69-9
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e4010c3c19cb174918df926fc7f07b1d722ad5a&vid=9e4010c3c19cb174918df926fc7f07b1d722ad5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR