2-Ethylaniline ≥98.0%

Supplier: TCI America
Warning

E0060-500ML E0060-25ML
TCE0060-500MLEA 147 USD
TCE0060-500ML TCE0060-25ML
2-Ethylaniline ≥98.0%
2-Ethylaniline

CAS Number: 578-54-1 MDL Number: MFCD00007751 Molecular Formula: C8H11N Molecular Weight: 121.18 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 216 Melting point (°C): -44 Flash Point (°C): 99 Specific Gravity (20/20): 0.98

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 210
Melting Pt: -46
Density: 0.98 (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00007751
CAS Number: 578-54-1
EINECS: 209-424-2
UN: 2273
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72754d773abed601d407f805ac0a64f5a9cc60f2&vid=833b45464acbcdb20105bece3077d12c4bdcaa29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR