2-Ethoxyethanol ≥99.0%

Supplier: TCI America
Danger

E0047-500ML E0047-25ML
TCE0047-500MLEA 44.32 USD
TCE0047-500ML TCE0047-25ML
2-Ethoxyethanol ≥99.0%
2-Ethoxyethanol

CAS Number: 110-80-5 MDL Number: MFCD00002869 Molecular Formula: C4H10O2 Molecular Weight: 90.12 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 135 Flash Point (°C): 44 Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: Cellosolve, Ethyl Cellosolve, Ethylene Glycol Monoethyl Ether, Ethyl Glycol

Formula: C₂H₅OCH₂CH₂OH
Boiling Pt: 135 °C
Melting Pt: –70 °C
Density: 0.93 (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
REACH: 01-2119560582-38
Merck Index: 13,03786
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54f06b5bc01b7cb47cc8e9f1e4721fec32a25ca&vid=f54f06b5bc01b7cb47cc8e9f1e4721fec32a25ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR