Sodium heptanoate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 10051-45-3
MDL Number: MFCD00070499
Molecular Formula: C7H14O2
Molecular Weight: 152.17
Purity/Analysis Method:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Enanthic Acid Sodium Salt, Sodium Enanthate, Heptanoic Acid Sodium Salt
Formula:
C₇H₁₄O₂ |
MDL Number:
MFCD00070499 CAS Number: 10051-45-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...