Dextrin, Soluble fiber
Supplier: TCI America
Synonyms:
Dextrin
D5658-100G
D5658-500G
TCD5658-100GEA
30
USD
TCD5658-100G
TCD5658-500G
Dextrin, Soluble fiber
Dextrin
Formula:
(C₆H₁₂O₆)n |
MDL Number:
MFCD00081554 CAS Number: 9004-53-9 EINECS: 232-675-4 Merck Index: 14,02953 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Solubility in hot Water | transparency |
Drying loss | max. 10.000 % |
Ignition residue (Sulfate) | max. 0.5 % |
Reducing suger | max. 8.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...