Dextran sulfate sodium salt, M.W. ~8000 (from dextran)

Supplier: TCI America

D5143-25G
TCD5143-25GEA 528 USD
TCD5143-25G
Dextran sulfate sodium salt, M.W. ~8000 (from dextran)
Dextran sulphate sodium salt
Formula: (C₆H₇Na₃O₁₄S₃)n
MDL Number: MFCD00081551
CAS Number: 9011-18-1
Merck Index: 14,02951

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Sulfur 15.0 to 20.0% (calcd.on dried substance)
Specific rotation [a]20/D +75 to +105 deg (C=5, H2O,calcd.on dried substance)
limiting viscosity 0.018 to 0.032
Drying loss max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR