3,6-Dimethoxy-9H-carbazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4833-1G
D4833-200MG
TCD4833-1GEA
247.9
USD
TCD4833-1G
TCD4833-200MG
3,6-Dimethoxy-9H-carbazole ≥98.0% (by GC)
3,6-Dimethoxy-9H-carbazole
Formula:
C₁₄H₁₃NO₂ |
MDL Number:
MFCD00626046 CAS Number: 57103-01-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...