4,5-Dimethoxy-2-nitrobenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

D4811-1G D4811-5G
TCD4811-1GEA 60 USD
TCD4811-1G TCD4811-5G
4,5-Dimethoxy-2-nitrobenzyl alcohol ≥98.0% (by GC)
4,5-Dimethoxy-2-nitrobenzyl alcohol
Formula: C₉H₁₁NO₅
MDL Number: MFCD00014701
CAS Number: 1016-58-6
EINECS: 213-806-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR