2,7-Diamino-9,9-di-n-octylfluorene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D4719-1G
D4719-200MG
TCD4719-1GEA
479.1
USD
TCD4719-1G
TCD4719-200MG
2,7-Diamino-9,9-di-n-octylfluorene ≥98.0% (by HPLC, titration analysis)
2,7-Diamino-9,9-di-n-octylfluorene
CAS Number: 851042-10-9
Molecular Formula: C29H44N2
Molecular Weight: 420.69
Purity/Analysis Method:
Formula:
C₂₉H₄₄N₂ |
MDL Number:
MFCD27923070 CAS Number: 851042-10-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...