2,6-Di(1-pyrazolyl)pyridine ≥98.0% (by GC)
Supplier: TCI America
D4672-1G
D4672-5G
TCD4672-1GEA
160.1
USD
TCD4672-1G
TCD4672-5G
2,6-Di(1-pyrazolyl)pyridine ≥98.0% (by GC)
2,6-Di(1-pyrazolyl)pyridine
CAS Number: 123640-38-0
Molecular Formula: C11H9N5
Molecular Weight: 211.23
Purity/Analysis Method:
Formula:
C₁₁H₉N₅ Melting Pt: 139 °C |
MDL Number:
MFCD28336856 CAS Number: 123640-38-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...