2,6-Dichloropyridine-4-methanol ≥98.0%
Supplier: TCI America
Synonyms:
2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine
D4423-1G
D4423-5G
TCD4423-1GEA
56.5
USD
TCD4423-1G
TCD4423-5G
2,6-Dichloropyridine-4-methanol ≥98.0%
2,6-Dichloropyridine-4-methanol
CAS Number: 101990-69-6
MDL Number: MFCD00052638
Molecular Formula: C6H5Cl2NO
Molecular Weight: 178.01
Purity/Analysis Method:
Other Product Names / Synonyms: 2,6-Dichloro-4-(hydroxymethyl)pyridine
Formula:
C₆H₅Cl₂NO Melting Pt: 136 °C |
MDL Number:
MFCD00052638 CAS Number: 101990-69-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...