2-(Diphenylphosphino)benzaldehyde ≥97.0%
Supplier: TCI America
Synonyms:
(2-Formylphenyl)diphenylphosphine
D4411-1G
D4411-5G
TCD4411-1GEA
42
USD
TCD4411-1G
TCD4411-5G
2-(Diphenylphosphino)benzaldehyde ≥97.0%
2-(Diphenylphosphino)benzaldehyde
CAS Number: 50777-76-9
MDL Number: MFCD00013367
Molecular Formula: C19H15OP
Molecular Weight: 290.30
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: (2-Formylphenyl)diphenylphosphine
Formula:
C₁₉H₁₅OP Melting Pt: 117 °C |
MDL Number:
MFCD00013367 CAS Number: 50777-76-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...