5,6-Dichlorobenzimidazole Hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D4295-1G
D4295-5G
TCD4295-1GEA
108.8
USD
TCD4295-1G
TCD4295-5G
5,6-Dichlorobenzimidazole Hydrochloride ≥98.0% (by HPLC, titration analysis)
5,6-Dichlorobenzimidazole Hydrochloride
CAS Number: 1087737-96-9
MDL Number: MFCD16810292
Molecular Formula: C7H4Cl2N2
Molecular Weight: 223.48
Purity/Analysis Method:
Formula:
C₇H₄Cl₂N₂ |
MDL Number:
MFCD16810292 CAS Number: 1087737-96-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...