N,N'-Diacetylchitobiose ≥98.0%
Supplier: TCI America
Synonyms:
GlcNAc beta(1-4)GlcNAc, 2-Acetamido-2-deoxy-4-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-D-glucopyranose
CAS Number: 35061-50-8
MDL Number: MFCD00077715
Molecular Formula: C16H28N2O11
Molecular Weight: 424.40
Purity/Analysis Method:
Product Categories: Bioscience, Disaccharides
Other Product Names / Synonyms: 2-Acetamido-2-deoxy-4-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl)-D-glucopyranose, GlcNAc beta(1-4)GlcNAc
Formula:
C₁₆H₂₈N₂O₁₁ MW: 424.40 g/mol Melting Pt: 247 °C |
MDL Number:
MFCD00077715 CAS Number: 35061-50-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...