N,N'-Di-n-octyl-3,4,9,10-perylenetetracarboxylic diimide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
PTCDI-C8, 2,9-Dioctylanthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetraone
D4175-1G
TCD4175-1GEA
195
USD
TCD4175-1G
N,N'-Di-n-octyl-3,4,9,10-perylenetetracarboxylic diimide ≥98.0% (by total nitrogen basis)
N,N'-Di-n-octyl-3,4,9,10-perylenetetracarboxylic diimide
CAS Number: 78151-58-3
MDL Number: MFCD08276854
Molecular Formula: C40H42N2O4
Molecular Weight: 614.79
Purity/Analysis Method:
Product Categories: Materials Chemistry, Organic Transistor Materials, Solar Cell Research Reagents, Perylene Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: PTCDI-C8
Formula:
C₄₀H₄₂N₂O₄ MW: 614.79 g/mol |
MDL Number:
MFCD08276854 CAS Number: 78151-58-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...