Desogestrel ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
13-Ethyl-11-methylene-18,19-dinor-17alpha-pregn-4-en-20-yn-17-ol
CAS Number: 54024-22-5
MDL Number: MFCD00869346
Molecular Formula: C22H30O
Molecular Weight: 310.48
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Bioscience, Other Pharmaceutical Ingredients, Other Steroids
Other Product Names / Synonyms: 13-Ethyl-11-methylene-18,19-dinor-17alpha-pregn-4-en-20-yn-17-ol
Formula:
C₂₂H₃₀O MW: 310.48 g/mol Melting Pt: 112 °C |
MDL Number:
MFCD00869346 CAS Number: 54024-22-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...