4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one ≥98.0% (by GC)
Supplier: TCI America
D4157-25G
TCD4157-25GEA
109.86
USD
TCD4157-25G
4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one ≥98.0% (by GC)
4,5-Dichloro-2-n-octyl-4-isothiazolin-3-one
CAS Number: 64359-81-5
MDL Number: MFCD04034673
Molecular Formula: C11H17Cl2NOS
Molecular Weight: 282.22
Purity/Analysis Method:
Formula:
C₁₁H₁₇Cl₂NOS Melting Pt: 42 °C |
MDL Number:
MFCD04034673 CAS Number: 64359-81-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...