Direct red 80 for biochemical research

Supplier: TCI America

D4132-5G
TCD4132-5GEA 34 USD
TCD4132-5G
Direct red 80 for biochemical research
Direct Red 80
Formula: C₄₅H₂₆N₁₀Na₆O₂₁S₆
MDL Number: MFCD00054389
CAS Number: 2610-10-8
EINECS: 220-027-3
Merck Index: 14,08548

Order Now

Specification Test Results

Appearance Red to Dark blue to Black powder to crystal
Lambda max. 524.0 to 528.0 nm (H2O)
Absorbance (E1%1cm) min. 350 (H2O, 524.0 to 528.0 nm)
Collagen detection test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR